Assay Details
| Gene Symbol: ldb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131314.1 | NP_571389.1 | 3 - 4 | 676 | 75 | |
| XM_005157342.4 | XP_005157399.3 | 3 - 4 | 571 | 75 | ||
| GenBank mRNA | AF031376.1 | - | 3 - 4 | 676 | 75 | |
| BC076070.1 | - | 3 - 4 | 508 | 75 | ||
More Information
Set Membership: |
|

ldb2a
Genomic Map