Assay Details
| Gene Symbol: npy4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131440.1 | NP_571515.1 | 1 | 1045 | 68 | |
| XM_005158664.4 | XP_005158721.1 | 2 | 1198 | 68 | ||
| GenBank mRNA | BC163146.1 | - | 1 | 1141 | 68 | |
| BC163152.1 | - | 1 | 1141 | 68 | ||
More Information
Set Membership: |
|

npy4r
Genomic Map