Assay Details
| Gene Symbol: dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131775.1 | NP_571850.1 | 4 - 5 | 456 | 94 | |
| GenBank mRNA | AY026507.1 | - | 4 - 5 | 376 | 94 | |
| BC071330.1 | - | 4 - 5 | 456 | 94 | ||
| EU145591.1 | - | 4 - 5 | 373 | 94 | ||
More Information
Set Membership: |
|

dhfr
Genomic Map