Assay Details
| Gene Symbol: cry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131787.1 | NP_571862.1 | 1 - 2 | 414 | 115 | |
| XM_005168277.4 | XP_005168334.1 | 2 - 3 | 641 | 115 | ||
| XM_009295766.3 | XP_009294041.1 | 2 - 3 | 628 | 115 | ||
| GenBank mRNA | AB042253.1 | - | 1 - 2 | 414 | 115 | |
| BC044385.1 | - | 2 - 3 | 548 | 115 | ||
| BC097079.1 | - | 2 - 3 | 578 | 115 | ||
| BC164413.1 | - | 2 - 3 | 511 | 115 | ||
More Information
Set Membership: |
|

cry4
Genomic Map