Assay Details
| Gene Symbol: cry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131787.1 | NP_571862.1 | 7 - 8 | 1269 | 92 | |
| GenBank mRNA | AB042253.1 | - | 7 - 8 | 1269 | 92 | |
| BC044385.1 | - | 8 - 9 | 1403 | 92 | ||
| BC164413.1 | - | 8 - 9 | 1366 | 92 | ||
More Information
Set Membership: |
|

cry4
Genomic Map