Assay Details
| Gene Symbol: aldh1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131850.1 | NP_571925.1 | 7 - 8 | 866 | 59 | |
| GenBank mRNA | AF288764.2 | - | 7 - 8 | 843 | 59 | |
| AF315691.1 | - | 7 - 8 | 854 | 59 | ||
| AF339837.1 | - | 7 - 8 | 866 | 59 | ||
| BC045851.1 | - | 7 - 8 | 855 | 59 | ||
| BC163897.1 | - | 7 - 8 | 829 | 59 | ||
More Information
Set Membership: |
|

aldh1a2
Genomic Map