Assay Details
| Gene Symbol: gpib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144764.2 | NP_658910.2 | 1 - 2 | 222 | 144 | |
| GenBank mRNA | AJ306396.1 | - | 1 - 2 | 225 | 144 | |
| BC121194.1 | - | 1 - 2 | 166 | 144 | ||
| EE689256.1 | - | 1 - 2 | 229 | 144 | ||
More Information
Set Membership: |
|

gpib
Genomic Map