Assay Details
| Gene Symbol: gpib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144764.2 | NP_658910.2 | 3 - 4 | 381 | 76 | |
| GenBank mRNA | AJ306396.1 | - | 3 - 4 | 384 | 76 | |
| BC121194.1 | - | 3 - 4 | 325 | 76 | ||
| EE689256.1 | - | 3 - 4 | 388 | 76 | ||
More Information
Set Membership: |
|

gpib
Genomic Map