Assay Details
Gene Symbol: gpib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144764.2 | NP_658910.2 | 4 - 5 | 505 | 83 | |
GenBank mRNA | AJ306396.1 | - | 4 - 5 | 508 | 83 | |
BC121194.1 | - | 4 - 5 | 449 | 83 | ||
EE689256.1 | - | 4 - 5 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
