Assay Details
| Gene Symbol: tp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152248.1 | NP_689454.1 | 8 - 9 | 1041 | 69 | |
| NM_152986.1 | NP_694518.1 | 8 - 9 | 1176 | 69 | ||
| NM_152987.1 | NP_694519.1 | 8 - 9 | 1193 | 69 | ||
| GenBank mRNA | AF412283.1 | - | 8 - 9 | 1176 | 69 | |
| AF412284.1 | - | 8 - 9 | 1041 | 69 | ||
| AF412285.1 | - | 8 - 9 | 1193 | 69 | ||
| AF487944.1 | - | 8 - 9 | 1041 | 69 | ||
More Information
Set Membership: |
|

tp63
Genomic Map