Assay Details
Gene Symbol: spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153663.2 | NP_705949.2 | 8 - 9 | 1159 | 74 | |
XM_005163832.4 | XP_005163889.1 | 8 - 9 | 1173 | 74 | ||
GenBank mRNA | AF465772.1 | - | 8 - 9 | 1115 | 74 | |
BC048024.1 | - | 8 - 9 | 1114 | 74 | ||
BC165625.1 | - | 7 - 8 | 959 | 74 | ||
CK027241.1 | - | 3 - 4 | 369 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
