Assay Details
| Gene Symbol: spns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153663.2 | NP_705949.2 | 9 - 10 | 1352 | 139 | |
| XM_005163832.4 | XP_005163889.1 | 9 - 10 | 1366 | 139 | ||
| GenBank mRNA | AF465772.1 | - | 9 - 10 | 1308 | 139 | |
| BC048024.1 | - | 9 - 10 | 1307 | 139 | ||
| BC165625.1 | - | 8 - 9 | 1152 | 139 | ||
| CF999294.1 | - | 1 - 2 | 200 | 139 | ||
| CK027241.1 | - | 4 - 5 | 562 | 139 | ||
More Information
Set Membership: |
|

spns1
Genomic Map