Assay Details
| Gene Symbol: tpi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153667.2 | NP_705953.1 | 6 - 7 | 700 | 72 | |
| GenBank mRNA | AF387820.1 | - | 6 - 7 | 700 | 72 | |
| BC049500.1 | - | 6 - 7 | 731 | 72 | ||
More Information
Set Membership: |
|

tpi1a
Genomic Map