Assay Details
| Gene Symbol: tpi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153668.4 | NP_705954.2 | 4 - 5 | 566 | 100 | |
| GenBank mRNA | AF387819.1 | - | 4 - 5 | 528 | 100 | |
| BC053294.1 | - | 4 - 5 | 506 | 100 | ||
| BC152271.1 | - | 4 - 5 | 518 | 100 | ||
| EH586742.1 | - | 4 - 5 | 572 | 100 | ||
More Information
Set Membership: |
|

tpi1b
Genomic Map