Assay Details
Gene Symbol: tpi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153668.4 | NP_705954.2 | 5 - 6 | 659 | 68 | |
GenBank mRNA | AF387819.1 | - | 5 - 6 | 621 | 68 | |
BC053294.1 | - | 5 - 6 | 68 | |||
BC152271.1 | - | 5 - 6 | 68 | |||
EH586742.1 | - | 5 - 6 | 665 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
