Assay Details
Gene Symbol: mknk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194402.1 | NP_919383.1 | 11 - 12 | 1174 | 64 | |
GenBank mRNA | BC044375.1 | - | 11 - 12 | 1174 | 64 | |
BC165746.1 | - | 10 - 11 | 978 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
