Assay Details
Gene Symbol: mknk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194402.1 | NP_919383.1 | 13 - 14 | 1384 | 71 | |
GenBank mRNA | BC044375.1 | - | 13 - 14 | 1384 | 71 | |
BC165746.1 | - | 12 - 13 | 1188 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
