Assay Details
| Gene Symbol: rbpja | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198878.2 | NP_942579.2 | 5 - 6 | 522 | 111 | |
| GenBank mRNA | AY128532.1 | - | 5 - 6 | 522 | 111 | |
| AY332617.1 | - | 5 - 6 | 495 | 111 | ||
| BC092918.1 | - | 5 - 6 | 499 | 111 | ||
| BC163922.1 | - | 5 - 6 | 522 | 111 | ||
More Information
Set Membership: |
|

rbpja
Genomic Map