Assay Details
| Gene Symbol: rbpja | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198878.2 | NP_942579.2 | 6 - 7 | 659 | 76 | |
| GenBank mRNA | AY128532.1 | - | 6 - 7 | 659 | 76 | |
| BC163922.1 | - | 6 - 7 | 659 | 76 | ||
More Information
Set Membership: |
|

rbpja
Genomic Map