Assay Details
| Gene Symbol: glud1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY577003.1 | - | 11 - 12 | 1658 | 64 | |
| BC044202.1 | - | 11 - 12 | 1594 | 64 | ||
| BC063940.1 | - | 11 - 12 | 1522 | 64 | ||
| BC165763.1 | - | 11 - 12 | 1476 | 64 | ||
More Information
Set Membership: |
|

glud1b
Genomic Map