Assay Details
| Gene Symbol: chtopa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199546.1 | NP_955840.1 | 3 - 4 | 521 | 93 | |
| XM_021467993.1 | XP_021323668.1 | 3 - 4 | 1154 | 93 | ||
| GenBank mRNA | BC045295.1 | - | 3 - 4 | 521 | 93 | |
More Information
Set Membership: |
|

chtopa
Genomic Map