Assay Details
| Gene Symbol: chtopa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199546.1 | NP_955840.1 | 4 - 5 | 668 | 89 | |
| XM_021467993.1 | XP_021323668.1 | 4 - 5 | 1301 | 89 | ||
| GenBank mRNA | BC045295.1 | - | 4 - 5 | 668 | 89 | |
More Information
Set Membership: |
|

chtopa
Genomic Map