Assay Details
| Gene Symbol: chtopa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199546.1 | NP_955840.1 | 5 - 6 | 805 | 56 | |
| XM_021467993.1 | XP_021323668.1 | 5 - 6 | 1438 | 56 | ||
| GenBank mRNA | BC045295.1 | - | 5 - 6 | 805 | 56 | |
More Information
Set Membership: |
|

chtopa
Genomic Map