Assay Details
Gene Symbol: waca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199660.1 | NP_955954.1 | 9 - 10 | 1243 | 70 | |
XM_005156197.2 | XP_005156254.1 | 9 - 10 | 1339 | 70 | ||
XM_009306588.2 | XP_009304863.1 | 9 - 10 | 1339 | 70 | ||
GenBank mRNA | AY576994.1 | - | 9 - 10 | 1248 | 70 | |
BC048888.1 | - | 9 - 10 | 1243 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
