Assay Details
| Gene Symbol: rps6kal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200073.1 | NP_956367.1 | 9 - 10 | 881 | 86 | |
| XM_009307718.2 | XP_009305993.1 | 4 - 5 | 378 | 86 | ||
| XM_021480984.1 | XP_021336659.1 | 2 - 3 | 186 | 86 | ||
| GenBank mRNA | BC045478.1 | - | 9 - 10 | 986 | 86 | |
| BC057467.1 | - | 9 - 10 | 881 | 86 | ||
More Information
Set Membership: |
|

rps6kal
Genomic Map