Assay Details
| Gene Symbol: kdelr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200103.1 | NP_956397.1 | 4 - 5 | 721 | 89 | |
| GenBank mRNA | BC056773.1 | - | 4 - 5 | 686 | 89 | |
| BC058065.1 | - | 4 - 5 | 721 | 89 | ||
More Information
Set Membership: |
|

kdelr2a
Genomic Map