Assay Details
| Gene Symbol: maco1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200319.2 | NP_956613.2 | 3 - 4 | 491 | 63 | |
| GenBank mRNA | AY845029.1 | - | 3 - 4 | 491 | 63 | |
| AY845031.1 | - | 3 - 4 | 517 | 63 | ||
| BC051623.1 | - | 3 - 4 | 491 | 63 | ||
More Information
Set Membership: |
|

maco1a
Genomic Map