Assay Details
| Gene Symbol: calb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200711.1 | NP_957005.1 | 8 - 9 | 633 | 115 | |
| GenBank mRNA | BC059467.1 | - | 8 - 9 | 633 | 115 | |
| FJ915062.1 | - | 3 - 4 | 149 | 115 | ||
More Information
Set Membership: |
|

calb2b
Genomic Map