Assay Details
| Gene Symbol: rps19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200750.1 | NP_957044.1 | 3 - 4 | 370 | 81 | |
| GenBank mRNA | BC059557.1 | - | 3 - 4 | 370 | 81 | |
| BC150459.1 | - | 4 - 5 | 387 | 81 | ||
More Information
Set Membership: |
|

rps19
Genomic Map