Assay Details
| Gene Symbol: hdac9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200816.2 | NP_957110.2 | 5 - 6 | 759 | 61 | |
| XM_005157964.3 | XP_005158021.1 | 5 - 6 | 1231 | 61 | ||
| GenBank mRNA | AY423001.1 | - | 5 - 6 | 743 | 61 | |
| BC059696.1 | - | 5 - 6 | 759 | 61 | ||
More Information
Set Membership: |
|

hdac9b
Genomic Map