Assay Details
| Gene Symbol: alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201287.1 | NP_958444.1 | 2 - 3 | 502 | 66 | |
| GenBank mRNA | BC054574.1 | - | 2 - 3 | 502 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alas1| Gene Symbol: alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201287.1 | NP_958444.1 | 2 - 3 | 502 | 66 | |
| GenBank mRNA | BC054574.1 | - | 2 - 3 | 502 | 66 | |
Set Membership: |
|