Assay Details
| Gene Symbol: rbm48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205604.2 | NP_991167.2 | 1 - 2 | 195 | 108 | |
| GenBank mRNA | AY423031.1 | - | 1 - 2 | 138 | 108 | |
| BC091857.1 | - | 1 - 2 | 141 | 108 | ||
| BC134007.1 | - | 1 - 2 | 137 | 108 | ||
| EH551204.1 | - | 1 - 2 | 178 | 108 | ||
More Information
Set Membership: |
|

rbm48
Genomic Map