Assay Details
| Gene Symbol: nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205729.2 | NP_991292.2 | 1 - 2 | 336 | 70 | |
| GenBank mRNA | AY391444.1 | - | 1 - 2 | 137 | 70 | |
| BC095127.1 | - | 1 - 2 | 305 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095127.1 transcripts.
Set Membership: |
|

nr1d1
Genomic Map