Assay Details
| Gene Symbol: cd9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212619.1 | NP_997784.1 | 3 - 4 | 271 | 73 | |
| XM_005158981.4 | XP_005159038.1 | 3 - 4 | 793 | 73 | ||
| GenBank mRNA | BC059691.1 | - | 3 - 4 | 384 | 73 | |
| BC067597.1 | - | 3 - 4 | 271 | 73 | ||
More Information
Set Membership: |
|

cd9a
Genomic Map