Assay Details
| Gene Symbol: eno1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212722.1 | NP_997887.1 | 5 - 6 | 378 | 113 | |
| XM_005162152.4 | XP_005162209.1 | 5 - 6 | 452 | 113 | ||
| GenBank mRNA | BC059511.1 | - | 5 - 6 | 378 | 113 | |
| BC071359.1 | - | 5 - 6 | 378 | 113 | ||
More Information
Set Membership: |
|

eno1a
Genomic Map