Assay Details
| Gene Symbol: hsd3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212797.1 | NP_997962.1 | 2 - 3 | 285 | 115 | |
| GenBank mRNA | AY279108.1 | - | 2 - 3 | 285 | 115 | |
| BC093118.1 | - | 2 - 3 | 274 | 115 | ||
| BC108022.1 | - | 2 - 3 | 288 | 115 | ||
| BC164774.1 | - | 1 - 2 | 189 | 115 | ||
More Information
Set Membership: |
|

hsd3b2
Genomic Map