Assay Details
| Gene Symbol: gnb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213202.1 | NP_998367.1 | 4 - 5 | 446 | 125 | |
| XM_005159345.4 | XP_005159402.1 | 4 - 5 | 455 | 125 | ||
| GenBank mRNA | BC059436.1 | - | 4 - 5 | 427 | 125 | |
| BC065866.1 | - | 4 - 5 | 446 | 125 | ||
More Information
Set Membership: |
|

gnb3b
Genomic Map