Assay Details
Gene Symbol: msmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213353.1 | NP_998518.1 | 5 - 6 | 762 | 95 | |
GenBank mRNA | BC050163.1 | - | 5 - 6 | 762 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
