Assay Details
| Gene Symbol: eif4bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214718.1 | NP_999883.1 | 7 - 8 | 868 | 82 | |
| XM_005166014.4 | XP_005166071.1 | 7 - 8 | 890 | 82 | ||
| XM_021476791.1 | XP_021332466.1 | 7 - 8 | 889 | 82 | ||
| GenBank mRNA | AY576997.1 | - | 7 - 8 | 868 | 82 | |
| BC066675.1 | - | 6 - 7 | 845 | 82 | ||
| BC086699.1 | - | 7 - 8 | 752 | 82 | ||
| BC164129.1 | - | 6 - 7 | 779 | 82 | ||
More Information
Set Membership: |
|

eif4bb
Genomic Map