Assay Details
Gene Symbol: nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002173.1 | NP_001002173.1 | 4 - 5 | 1165 | 71 | |
GenBank mRNA | AY542372.2 | - | 4 - 5 | 1162 | 71 | |
BC074092.1 | - | 4 - 5 | 1165 | 71 | ||
BC164696.1 | - | 3 - 4 | 1121 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
