Assay Details
| Gene Symbol: twf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002304.1 | NP_001002304.1 | 6 - 7 | 660 | 78 | |
| GenBank mRNA | BC074070.1 | - | 6 - 7 | 660 | 78 | |
| BC164001.1 | - | 6 - 7 | 646 | 78 | ||
More Information
Set Membership: |
|

twf1b
Genomic Map