Assay Details
Gene Symbol: twf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002304.1 | NP_001002304.1 | 7 - 8 | 804 | 122 | |
GenBank mRNA | BC074070.1 | - | 7 - 8 | 804 | 122 | |
BC164001.1 | - | 7 - 8 | 790 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
