Assay Details
| Gene Symbol: glrx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002404.2 | NP_001002404.1 | 2 - 3 | 211 | 116 | |
| XM_005161733.4 | XP_005161790.1 | 2 - 3 | 269 | 116 | ||
| XM_005161734.4 | XP_005161791.1 | 2 - 3 | 145 | 116 | ||
| XM_005161735.3 | XP_005161792.1 | 2 - 3 | 231 | 116 | ||
| XM_009296369.3 | XP_009294644.1 | 2 - 3 | 80 | 116 | ||
| GenBank mRNA | BC076178.1 | - | 2 - 3 | 212 | 116 | |
More Information
Set Membership: |
|

glrx2
Genomic Map