Assay Details
| Gene Symbol: dhrs3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003477.1 | NP_001003477.1 | 3 - 4 | 635 | 59 | |
| GenBank mRNA | BC078383.1 | - | 3 - 4 | 635 | 59 | |
| BC164069.1 | - | 3 - 4 | 494 | 59 | ||
More Information
Set Membership: |
|

dhrs3a
Genomic Map