Assay Details
| Gene Symbol: gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003491.2 | NP_001003491.2 | 9 - 10 | 1166 | 63 | |
| GenBank mRNA | BC078357.1 | - | 9 - 10 | 1130 | 63 | |
| EH459927.1 | - | 1 - 2 | 682 | 63 | ||
| EH541442.1 | - | 5 - 6 | 708 | 63 | ||
| FJ392634.1 | - | 1 - 2 | 88 | 63 | ||
More Information
Set Membership: |
|

gmppb
Genomic Map