Assay Details
| Gene Symbol: st6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003853.1 | NP_001003853.1 | 1 - 2 | 194 | 59 | |
| XM_005161298.4 | XP_005161355.1 | 2 - 3 | 834 | 59 | ||
| XM_005161299.4 | XP_005161356.1 | 2 - 3 | 274 | 59 | ||
| GenBank mRNA | AJ744801.1 | - | 1 - 2 | 194 | 59 | |
| BC095555.1 | - | 2 - 3 | 280 | 59 | ||
| BC117603.1 | - | 2 - 3 | 255 | 59 | ||
More Information
Set Membership: |
|

st6gal1
Genomic Map