Assay Details
| Gene Symbol: rcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003865.1 | NP_001003865.1 | 2 - 3 | 211 | 81 | |
| XM_021476091.1 | XP_021331766.1 | 2 - 3 | 344 | 81 | ||
| GenBank mRNA | AY648776.1 | - | 2 - 3 | 211 | 81 | |
| BC095051.1 | - | 2 - 3 | 272 | 81 | ||
| BC164769.1 | - | 2 - 3 | 216 | 81 | ||
More Information
Set Membership: |
|

rcl1
Genomic Map