Assay Details
Gene Symbol: myo6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004110.1 | NP_001004110.1 | 28 - 29 | 3122 | 73 | |
XM_021467147.1 | XP_021322822.1 | 29 - 30 | 3380 | 73 | ||
XM_021467148.1 | XP_021322823.1 | 28 - 29 | 3370 | 73 | ||
GenBank mRNA | AY618701.1 | - | 28 - 29 | 3122 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
