Assay Details
Gene Symbol: myo6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004110.1 | NP_001004110.1 | 2 - 3 | 183 | 71 | |
XM_005158836.4 | XP_005158893.1 | 3 - 4 | 437 | 71 | ||
XM_009293188.3 | XP_009291463.1 | 3 - 4 | 437 | 71 | ||
XM_021467147.1 | XP_021322822.1 | 3 - 4 | 441 | 71 | ||
XM_021467148.1 | XP_021322823.1 | 3 - 4 | 440 | 71 | ||
GenBank mRNA | AY618701.1 | - | 2 - 3 | 183 | 71 | |
AY691328.1 | - | 3 - 4 | 336 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
