Assay Details
| Gene Symbol: myo6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004110.1 | NP_001004110.1 | 3 - 4 | 265 | 66 | |
| XM_005158836.4 | XP_005158893.1 | 4 - 5 | 519 | 66 | ||
| XM_009293188.3 | XP_009291463.1 | 4 - 5 | 519 | 66 | ||
| XM_021467147.1 | XP_021322822.1 | 4 - 5 | 523 | 66 | ||
| XM_021467148.1 | XP_021322823.1 | 4 - 5 | 522 | 66 | ||
| GenBank mRNA | AY618701.1 | - | 3 - 4 | 265 | 66 | |
| AY691328.1 | - | 4 - 5 | 418 | 66 | ||
More Information
Set Membership: |
|

myo6b
Genomic Map